Learn More
Poltex PETG Short Glove Box Holder
Description
- Constructed from extremely durable, clear PETG plastic
- Lift-off face and short stature allows for mounting in compact spaces
- Front face hooks on with a secure hold, but easily lifts off to replace empty glove boxes
- Mounting hardware is not included for keyhole mount model
Specifications
Specifications
| Product Type | Glove Box Holder |
| Color | Clear |
| Dimensions (D x W x H) Interior | 3.7 x 11 x 7 in. |
| Holds | Disposable Glove Boxes |
| Material | PETG |
| Mounting | Keyhole Slots |
| No. of Compartments | 2 |
| Product Style | Top Loading |
| Capacity | 2 Boxes |