Learn More
Dräger™ Short-Term Detector Tubes: Chlorine
Catalog No. 17985206 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
0.2 to 30
50 to 500
Tube Type:
Chlorine 0.2/a
Chlorine 50/a
Humidity Range:
Less than40mg/L
Not Applicable
No. of Strokes:
1
10/1
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Tube Type Humidity Range No. of Strokes
17-985-206 0.2 to 30 Chlorine 0.2/a Not Applicable 10/1
17-985-207 50 to 500 Chlorine 50/a Less than40mg/L 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-985-206 Supplier Dräger™ Supplier No. CH24301
  • $213.00
    $171.65 / Pack of 10
    Save $41.35
    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Components Measured Chlorine
Type Gas Detection Tube
Humidity Range Not Applicable
Measurement Range 0.2 to 30
No. of Strokes 10/1
Quantity 10
Response Time 3 min./20 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
Tube Type Chlorine 0.2/a