Learn More
Description
- Features an anti-fog, anti-scratch coating.
- Available in clear, grey, and amber.
- Fits 3M™ SecureFit™ X5000 Series Safety Helmets
- 3M Catalog# X5-SV01 , 3M Catalog# X5-SV02 , 3M Catalog# X5-SV03
Specifications
Specifications
| Color | Amber |
| For Use With (Application) | Demolition, Forestry, Grinding, Remediation |
| Material | Polycarbonate |
| Model | X5-SV03 |
| Product Type | Short Visor |
| Quantity | 10/Cs. |
| Size | One Size Fits Most |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.