Learn More
MilliporeSigma™ Supelco™ Shortix™ Tubing Cutter
Catalog No. 111008787 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
1/Pk.
Catalog No. Packaging Quantity
11-100-8787 1/Pk.
1 options
Catalog No. 11-100-8787 Supplier MilliporeSigma™ Supelco™ Supplier No. 21386U
Add to Cart
Add to Cart
Correct fitting of a ferrule is very important in column performance.
The ferrule needs to be properly sized and squarely cut using a tubing cutter and the cut should be examined by a magnifier. This is important as column fragments or ferrule particles can contaminate the column. Hence, the column ends are pointed towards the floor during cutting.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 1/Pk.
Shortix is a trademark of Scientific Glass Technology B.V.