Learn More
SHOWA™ 540 HPPE Cut-Resistant Gloves, Polyurethane Coated
Catalog No. 191305613E Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-5613E 2X-Large
19-130-5613B Medium
19-130-5613C Large
19-130-5613D X-Large
19-130-5613A Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-130-5613E Supplier SHOWA™ Supplier No. 540XXL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Offers a combination of exceptional grip, dexterity and cut-resistance with the durability of a polyurethane coating.
HPPE lining provides comfortable fit
  • Durable polyurethane palm-coated glove
  • 13-gauge cut-resistant liner consisting of High Performance Polyethylene Engineered (HPPE)
  • Elastic knit-wrist secures the glove above the wrist to prevent debris from entering the glove
  • ANSI Cut Level A2
  • Launders at 40°C (104°F) using a neutral detergent
Specifications
ANSI Abrasion Rating ANSI 6
Certifications/Compliance (Europe) CE
Construction Seamless Knit
Coating Material Polyurethane
Color White
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free Yes
Material HPPE
Product Type Cut-resistant Gloves
Size 2X-Large
EN388 Standard EN 388:4241
For Use With (Application) Machining, scrap metal handling, glazing, glass handling, automotive industry, cutting fluids, construction, HVAC, electronic assembly, maintenance
Glove Cuff Style Knit
Model 540-XXL
Puncture Rating ANSI/ISEA 105-16 Level 2
Series 540 Series
Yarn Gauge 13
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.