Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 541 HPPE Cut-Resistant Gloves, Polyurethane Coating

Catalog No. 19823381 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-823-381 Large
19-270-221 Small
19-270-222 Medium
19-270-224 X-Large
19-270-225 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-823-381 Supplier SHOWA™ Supplier No. 541L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rough grip gloves provide a high degree of comfort and control when performing intricate tasks. Dark color hides grime in dirty environments.

  • Durable flat-dipped gray colored polyurethane-coated glove
  • 13-gauge HPPE cut-resistant liner
  • Elastic knit-wrist secures glove above wrist for comfort and prevents debris from entering glove

Specifications

Certifications/Compliance (Europe) Cat II CE
Construction Seamless Knit
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A3
Hand Specific
Latex Free Yes
Material HPPE
Product Type Cut-resistant Gloves
Size Large
EN388 Standard EN 388:4342B
For Use With (Application) Intricate sharp parts handling, construction, glass handling, bottling, canning, sheet metal work, automotive, fabrication
Glove Cuff Style Elasticated
Model 541-L
Puncture Rating ANSI/ISEA 105-16 Level 3
Series 541 Series
Length (English) 6 in.
Length (Metric) 15.2 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.