Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ AEGIS HP54 Cut-Resistant Gloves

Catalog No. 19120553 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-120-553 Large
19-120-551 Small
19-120-552 Medium
19-120-554 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-120-553 Supplier SHOWA™ Supplier No. 23009
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thin, lightweight seamless knit gloves are comprised of HPPE, nylon and lycra with Zorb-IT™ palm coating for a secure grip.

  • 10-gauge seamless knit comprised
  • High Performance Poly Ethylene (HPPE), nylon, lycra with Zorb-IT™ palm coating
  • Combination of comfort, cut resistance and durability
  • Thin, lightweight fit and feel

Specifications

Certifications/Compliance (Europe) Cat II, CE
Construction Seamless Knit
Coating Material Sponge Nitrile
Color Black, White
Cut Resistance Rating ANSI A4
Material HPPE, Nylon, Lycra
Product Type Cut-resistant Gloves
Size Large
EN388 Standard EN 388:4542
For Use With (Application) Oily surfaces, Metal stamping, mechanical and automotive, sheet Metal assembly, Glass handling, bottling, recycling
Model 230-09
Series AEGIS HP54
Yarn Gauge 10
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.