Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 460 Series PVC-Coated Cold Resistant Gloves

Catalog No. 19270238 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-270-238 Medium
19-019-247 Small
19-270-239 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-270-238 Supplier SHOWA™ Supplier No. 46008
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Insulated gloves protect hands, and remain flexible in low temperatures. Rough-textured PVC coating protects against oils and chemicals, while also enhancing grip capabilities and tactile precision.

  • Gloves remain flexible in temperatures as low as -4°F (-20°C)
  • PVC coating protects against oils and chemicals
  • Rough grip enhances tactile precision when handling small, greasy components
  • Extended wrist protection from harmful substances
  • Reduced potential exposure to bacteria, viruses, and fungi

Specifications

Coated Area Full
Coating Material PVC
Color Orange
Exterior Finish Fully Textured
Hand Specific
Size Medium
For Use With (Application) Chemical Spray/Treatment, Coating Preparation, and Painting
Glove Cuff Style Gauntlet
Liner Material Acrylic
Series 460
Length (English) 11.8 in.
Length (Metric) 30 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.