Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Triple-Dipped PVC Gloves with Aramid Fiber Lining

Catalog No. 19270248 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-270-248 X-Large
19-270-246 Medium
19-270-247 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-270-248 Supplier SHOWA™ Supplier No. KV66010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves provide protection against a wide range of hazards including cuts, abrasions, chemicals, and oils. Rough textured surface increases durability and enhances grip capabilities.

  • Triple dipped PVC coating protects against oils and chemicals
  • Kevlar™ aramid fiber liner provides cut resistance
  • Rough textured surface enhances grip capabilities
  • Hazard barrier is maintained even after multiple washes
  • ANSI/ISEA 105-16 Level 4 Puncture Resistance

Specifications

ANSI Abrasion Rating ANSI 6
Certifications/Compliance CE (Cat III); EN 388:2016+A1:2018 (4X42B); EN ISO 374-1:2016+A1:2018 Type JKL; EN ISO 374-5:2016 (Virus); EN 407:2020 (X1XXXX); EN ISO 21420:2020; ANSI/ISEA 105-2016 Cut Level A2; ANSI/ISEA 105-2016 Abrasion Level 6; ANSI/ISEA 105-2016 Puncture Level 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Coated Area Full Glove
Construction Seamless Knit
Coating Material PVC
Color Blue
Cut Resistance Rating ANSI A2
Exterior Finish Fully Textured
Hand Specific
Product Type Dip-coated Gloves
Size X-Large
EN388 Standard 4X42B
EN ISO 374-1 CHEMICAL LETTER CODES JKL
EN ISO 374 - 5 Bacteria, Fungi only
For Use With (Application) Chemical Spray, Coating Preparation, Drainage, HVAC, Plumbing, and Painting
Glove Cuff Style Open
Grip Design Rough
Liner Material Aramid Fiber
Puncture Rating ANSI/ISEA 105-16 Level 4
Resistant To Oils, Chemicals, Abrasion, Cuts, Puncture
Series KV660
Length (English) 12 in.
Length (Metric) 30.5 cm
Product Line KV660
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.