Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Ultimate N-DEX™ 9905 Nitrile Gloves
GREENER_CHOICE

Catalog No. 19120216D Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-120-216D X-Large
19-120-216A Small
19-120-216B Medium
19-120-216C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-120-216D Supplier SHOWA™ Supplier No. 9905PFXL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ultimate N-DEX™ Nitrile Gloves are engineered with extended-length and a thicker mil for tactile precision, maximum comfort and protection in a wide-range of applications.

  • Thicker mil provides added barrier protection
  • 11 in. longer cuff to protect wrists
  • Second skin fit and feel
  • Rolled cuff prevents dirt from entering the glove
  • Recommended for agricultural, food services, municipal services, pharmaceutical and utilities
  • Made in the USA

Specifications

Certifications/Compliance 21 CFR 170-199, ASTM
Applications Industrial
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish No Texture
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size X-Large
Thickness (English) 6 mil
Thickness (Metric) 0.15 mm
Model 9905PFXL
Length (English) 11 in.
Length (Metric) 28 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.