Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ N-DEX™ Accelerator-Free Nitrile Gloves

Catalog No. 19170357E Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-170-357E X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-170-357E Supplier SHOWA™ Supplier No. 7705PFTXL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Accelerator-free, highly visible disposable nitrile gloves are ideal for laboratory, pharmaceutical, or industrial tasks

 Ideal for jobs requiring high visibility, quick identification.

  • Nitrile construction
  • Durable, puncture-resistant
  • Powder free, ambidextrous
  • Textured finger
  • Color: High-vis Green

Specifications

Certifications/Compliance ASTM F 1671, ASTM D 6319, NFPA 1999, FDA 21 CFR 170-199, ISO 9002
Accelerator Free Yes
Applications Industrial
Color Green
Disposable Single-use
Exterior Finish Fingertip Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Gloves
Size X-Large
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
Model 7705PFTXL
Length (English) 9.5 in.
Length (Metric) 24.13 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.