Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ N-DEX™ 6005PF Disposable Nitrile Gloves

Catalog No. 191202415B Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
X-Small
Small
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-120-2415B Small
19-120-2415A X-Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-120-2415B Supplier SHOWA™ Supplier No. 6005PFSM

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

N-DEX™ 6005 series powder free gloves provide enhanced dexterity and performance across a range of tactical applications including food, electronics, janitorial, municipal services and utilities.

  • Easy to put on and remove
  • Can easily be one-hand used
  • Ambidextrous
  • Rolled cuff
  • Smooth grip
  • Made in the U.S.A.
  • Specific applications include food handling, agriculture, washing and cleaning, public utility and coroner/morgue

Specifications

Certifications/Compliance 21 CFR 170-199
Certifications/Compliance (Europe) EN 455, Cat. I
Applications Industrial
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish No Texture
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Small
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
Model 6005PFSM
Length (English) 9.5 in.
Length (Metric) 24 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.