Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 6784R Chemical Resistant Neoprene Gloves, Cotton Liner

Catalog No. 19019202 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-019202 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-019202 Supplier SHOWA™ Supplier No. 6784R10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical resistant neoprene glove with soft cotton liner features a rough coating to protect against acids, chemicals and caustics.

  • Chemical resistant against acids, caustics, oils, greases and many solvents
  • Protection against physical hazards such as abrasion and cut
  • Rough grip for wet applications
  • Cotton liner keeps cool and comfortable in hot conditions or warm in cold conditions
  • Ideal for biotechnology, cytostatic, life sciences, pharmaceuticals and API

Specifications

Certifications/Compliance 21 CFR 170-199
Certifications/Compliance (Europe) Cat III CE 0321, EN 407:4342XX
Applications Healthcare
Color Black
Latex Free Yes
Material Neoprene
Product Type Neoprene Gloves
Size 10
EN388 Standard EN 388:3111X
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AJKLMT
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Gauntlet
Grip Design Rough
Liner Material Cotton
Lining Lined
Model 6784R
Series 6784R
Length (English) 14 in.
Length (Metric) 35.5 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.