Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 7066 Smooth Nitrile Coated Cotton Oil & Grease Repellent Gloves

Catalog No. 1139513 Shop All Showa Best Glove Products
Change view
Click to view available options
Series:
7066
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Series
11-395-13 7066
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-395-13 Supplier SHOWA™ Supplier No. 706610
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Exclusive coating prevents oil breakthrough while protecting the palms from rough and abrasive materials. Smooth coating enhances grip in dry and oily conditions.

  • Exclusive coating provides cut, puncture, snag, and abrasion resistance
  • Smooth finish enhances grip in dry and oily conditions
  • Barrier is resistant to oil and grease, and prevents oil breakthrough
  • Reinforced safety cuff for additional protection
  • Economical replacement for leather

Specifications

Certifications/Compliance CE Category II, EN 388:2016+A1:2018 3111X
Certifications/Compliance (Europe) CE Category II, EN 388:2016+A1:2018 3111X
Coated Area Full Glove
Construction Cut and Sewn
Cuff Material Reinforced Cotton
Coating Material Nitrile
Color Navy
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Cotton
Product Type Dip-coated Gloves
Size Large
For Use With (Application) Mechanical Engineering, Light Chemical Handling, Landscaping, and Equipment Operation
Glove Cuff Color Natural
Glove Cuff Style Safety
Grip Design Smooth
Liner Material Cotton
Packaging Type Polybag
Resistant To Oil, Grease, Abrasion, Cuts, Punctures, Snags
Series 7066
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.