Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Shrimp Alkaline Phosphatase (SAP)

Catalog No. 783905000UN
Encompass_Preferred
Change view
Click to view available options
Quantity:
1000 Units
500 Units
5000 Units
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
78-390-5000UN 5000 Units
78-390-1000UN 1000 Units
78-390-500UN 500 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 78-390-5000UN Supplier Applied Biosystems™ Supplier No. 783905000UN
Add to Cart
Edge
Add to Cart

Shrimp Alkaline Phosphatase (SAP) is a high specific activity, heat-labile alkaline phosphatase purified from a recombinant source and originally isolated from Pandalus borealis (arctic shrimp).

  • 100% heat-inactivated in 5 min at 65°C
  • Significantly improved storage stability at lower temperatures
  • Very high specific activity
  • Removes 5'-phosphates from DNA, RNA, dNTPs, and proteins
  • Purified from a recombinant source
  • May be added directly to restriction enzyme digests
  • No vector purification necessary
  • Requires no supplemental zinc or other additives for activity
  • Works direct in many different buffers
  • Easy treatment of unincorporated dNTPs in PCR products prior to DNA sequencing or SNP analysis

Order Info

500 Units

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 unit/μL
Enzyme Alkaline Phosphatase
For Use With (Application) Dephosphorylation of DNA/RNA
Quantity 5000 Units

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.