Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Shutoff Valve, Low Pressure, Tefzel™™

Catalog No. 111004994 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With:
1/8 in. Tubing
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
11-100-4994 1/8 in. Tubing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-4994 Supplier MilliporeSigma™ Supelco™ Supplier No. 505773
Add to Cart
Add to Cart

Designed for lower pressure applications . Biocompatible, all polymer flow path. PEEK or Tefzel™ valve with 1/4-28 fittings, for pressures up to 1000 psi (70 kg/cm2). Flangeless nuts and ferrules included.

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Description Number of positions 2
For Use With 1/8 in. Tubing
Material Tefzel

Tefzel is a registered trademark of E. I. du Pont de Nemours and Company

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.