Learn More
SINGH INSTRUMENT LLC M 96 - High Throughput Pressure Reactor for SBS format Plates and Blocks
Supplier:  SINGH INSTRUMENT LLC SIFS1004
Encompass_Preferred
M 96 offers exceptional process control capabilities at an remarkably small footprint of 8.5 in width and 13 in deep. M 96 is easy to move in and out of a glovebox through an anti-chamber. M 96 can house SBS format plates and blocks, ranging from 96 well (1ml glass inserts) to 6 well (20ml glass vials). Equipped with two temperature zones this reactor has uniform heating across all reaction chambers. Temperature range is RT to 200C. High temperatures are possible. All units come standard with cooling channels. An external chiller can cool the reactor post reactions to minimize down time between experiments.M96 comes with a built-in digital pressure regulator with pressure range from ambient to 600 PSIG. Wetted materials are 316SS and Viton/FFKM. High pressures are possible, inquire for more options. M-96 has integrated stirrer – the same technology is used in LiStir 3P. The homogeneous mixing of this stirrer gives M96 added advantage.
Catalog No. 50-196-3634
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.