Learn More
Description
- Bright blue calculator is solar powered and battery powered
- Perfect for independent math learning
- It is four function calculator that has three key memory, auto shut off and an 8-digit display
Specifications
Specifications
| Product Type | Primary Calculator |
| Display Type | Digital |
| Color | Bright Blue |
| Key Functions | Addition, subtraction, multiplication and division |
| Memory | 3-key |
| Quantity | 1 Ea. |
| Power Supply | Battery, Solar |
| For Use With (Application) | Great tool for beginning to teach calculator skills |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
