Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SI Metric Manufacturing Primary Calculator
SCI_ED

Catalog No. S95604 Shop All Si Metric Manufacturing Limited Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S95604 10/Pk.
S95603 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S95604 Supplier Si Metric Manufacturing Limited Supplier No. 50240

missing translation for 'validMainframeMsg'

A great tool for beginning to teach calculator skills.

  • Bright blue calculator is solar powered and battery powered
  • Perfect for independent math learning
  • It is four function calculator that has three key memory, auto shut off and an 8-digit display

Specifications

Product Type Primary Calculator
Display Type Digital
Color Bright Blue
Key Functions Addition, subtraction, multiplication and division
Memory 3-key
Quantity 10/Pk.
Power Supply Battery, Solar
For Use With (Application) Great tool for beginning to teach calculator skills
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.