Description
Includes
CHO cell-expressed Recombinant Human CD73, CMP, Phosphatase Buffer 2, 100mM MnCl2, Phosphate Standard, Malachite Green Reagents A and B
Uses the 5 ft.-nucleotidase CD73 as a coupling phosphatase to remove inorganic phosphate quantitatively from the leaving nucleotide cytidine 5 ft.-monophosphate (CMP) that is generated during sialyltransferase reactions
No t
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Product Line | R&D Systems |
| Quantity | 1 Kit |