Learn More
ENG Scientific Siderocyte Stain Kit
Description
- Siderocyte Stain Kit
- Staining time 5 seconds
- For in-vitro diagnostic use only
Specifications
Specifications
| Quantity | 2 x 250 mL |
| For Use With (Application) | For in-vitro diagnostic use only |
| Includes | Solution II - Hydrochloric Acid (4%) |
| Packaging | Bottles |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.