Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIDT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534493 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534493 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534493 Supplier Thermo Scientific Supplier No. PA534493
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

SIDT2 Polyclonal Antibody

SIDT2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen SIDT2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SIDT2
Gene Accession No. D3ZEH5, Q8CIF6, Q8NBJ9
Gene Alias B930096O19; BC023957; CGI-40; PSEC0072; RGD1308311; SID1 transmembrane family member 2; SID1 transmembrane family, member 2; Sidt2; UNQ685/PRO1325
Gene Symbols SIDT2
Host Species Rabbit
Immunogen An 18 amino acid peptide near the amino terminus of human SIDT2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 214597, 315617, 51092
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.