Learn More
Siemens Healthineers Urinalysis Reagent Test Strips
Catalog No. AM2191 Shop All Siemens Healthineers Products
Change view
Click to view available options
Detectable Analytes:
Bilirubin, Blood, Glucose, Ketone, pH, Protein, Urobilinogen
Blood
Blood Urea, Nitrogen
Blood, Glucose, Ketone, pH, Protein
Blood, Glucose, pH, Protein
Glucose, Protein
Protein
Quantity:
100 Pack
25 Pack
50 Pack
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes Quantity
AM-2191 Protein 100 Pack
AM-2830 Blood Urea, Nitrogen 25 Pack
AM-2179 Bilirubin, Blood, Glucose, Ketone, pH, Protein, Urobilinogen 100 Pack
23-042320 Blood, Glucose, Ketone, pH, Protein 100 Pack
23-042321 Blood, Glucose, pH, Protein 100 Pack
AM-2190 Blood 50 Pack
AM-2184 Glucose, Protein 100 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. AM-2191 Supplier Siemens Healthineers Supplier No. 10333485
Add to Cart
Add to Cart
Single-use, plastic strips with multiple reagent test areas
  • Ready to use and entirely disposable
  • Dip and read visually for results
  • Some strips can be read instrumentally on Clinitek™ 50, 100, 200, 200+, 500, Advantus, and Status Family of Urine Chemistry analyzers
TRUSTED_SUSTAINABILITY
Specifications
Detectable Analytes Protein
Disposable Single-use
For Use With (Application) Urinalysis, in-vitro diagnostic use
Quantity 100 Pack