Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIGB Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144540
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144540 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144540 Supplier Invitrogen™ Supplier No. PA5144540
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SIGB Polyclonal Antibody for Western Blot, ELISA

Required for the transition of plastids into chloroplasts by coordinating nuclear and chloroplastic genomes under light conditions. Sigma factors are initiation factors that promote the attachment of plastid-encoded RNA polymerase (PEP) to specific initiation sites and are then released. Promotes the biosynthesis of plastid-encoded tRNAs (e.g. trnE-UUC and trnV-UAC).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIGB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.274 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene SIGB
Gene Accession No. O22056
Gene Alias ABC1; SIG1; SIG2; SIGA
Gene Symbols SIGB
Host Species Rabbit
Immunogen Recombinant Arabidopsis thaliana SIGB protein (40-310aa).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 837376
Target Species Plant
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.