Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIGLEC H Monoclonal Antibody (eBio440c), Functional Grade, eBioscience™
GREENER_CHOICE

Catalog No. p-7084939
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-142-91 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-142-91 Supplier Invitrogen™ Supplier No. 16033382
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The monoclonal antibody eBio440c recognizes Siglec-H, a protein exclusively found on pDC (plasmacytoid dendritic cells) or type I IFN-producing cells (IPC) in the naive mouse. mouse IPC are typically PDCA+, CD11c+, CD11b-, B220+, and Ly-6C+, and are quick to respond to viruses. Siglec-H is a transmembrane protein of the Ig superfamily that like CD33 have been shown to bind sialic acid but lacks the characteristic cytoplasmic ITIM domain (immunoreceptor tyrosine based inhibitory motif). To overcome the lack of a cytoplasmic domain, Siglec-H associates with DAP12 thereby allowing for signal transduction. The eBio440c antibody has been shown to inhibit pDC function (inhbits IFNalpha secretion in response to CpG). It has been observed that some mouse strains (such as SJL) have higher percentages of pDCs compared to C57BL/6. Applications Reported: This eBio440c antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio440c antibody has been tested by blocking staining of the direct conjugate of eBio440c. This can be used at less than or equal to 2.0 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. It is recommended that direct conjugates such as Product # 1...

Siglec-H, a protein exclusively found on pDC (plasmacytoid dendritic cells) or type I IFN-producing cells (IPC) in the naive mouse. Mouse IPC are typically PDCA+, CD11c+, CD11b-, B220+, and Ly-6C+, and are quick to respond to viruses. Siglec-H is a transmembrane protein of the Ig superfamily that like CD33 have been shown to bind sialic acid but lacks the characteristic cytoplasmic ITIM domain (immunoreceptor tyrosine based inhibitory motif). To overcome the lack of a cytoplasmic domain, Siglec-H associates with DAP12 thereby allowing for signal transduction. The eBio440c antibody has been shown to inhibit pDC function (inhbits IFNalpha secretion in response to CpG). It has been observed that some mouse strains (such as SJL) have higher percentages of pDCs compared to C57BL/6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIGLEC H
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
Clone eBio440c
Concentration 1 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2
Gene Siglech
Gene Accession No. 0
Gene Alias 6430529G09Rik; sialic acid binding Ig-like lectin H; Siglec; Siglech; Siglec-H; SIGLEC-like protein
Gene Symbols Siglech
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233274
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.