Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIGLEC H Monoclonal Antibody (eBio440c), Super Bright™ 436, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 62033382
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-033-382 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 62-033-382 Supplier Invitrogen™ Supplier No. 62033382
  • $484.00
    $363.65 / Each of 1

    Save $120.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

The monoclonal antibody eBio440c recognizes Siglec-H, a protein exclusively found on pDC (plasmacytoid dendritic cells) or type I IFN-producing cells (IPC) in the naive mouse. Mouse IPC are typically PDCA+, CD11c+, CD11b-, B220+, and Ly-6C+

Siglec-H, a protein exclusively found on pDC (plasmacytoid dendritic cells) or type I IFN-producing cells (IPC) in the naive mouse. Mouse IPC are typically PDCA+, CD11c+, CD11b-, B220+, and Ly-6C+, and are quick to respond to viruses. Siglec-H is a transmembrane protein of the Ig superfamily that like CD33 have been shown to bind sialic acid but lacks the characteristic cytoplasmic ITIM domain (immunoreceptor tyrosine based inhibitory motif). To overcome the lack of a cytoplasmic domain, Siglec-H associates with DAP12 thereby allowing for signal transduction. The eBio440c antibody has been shown to inhibit pDC function (inhbits IFNalpha secretion in response to CpG). It has been observed that some mouse strains (such as SJL) have higher percentages of pDCs compared to C57BL/6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIGLEC H
Applications Flow Cytometry
Classification Monoclonal
Clone eBio440c
Concentration 0.2 mg/mL
Conjugate Super Bright 436
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Siglech
Gene Accession No. 0
Gene Alias 6430529G09Rik; sialic acid binding Ig-like lectin H; Siglec; Siglech; Siglec-H; SIGLEC-like protein
Gene Symbols Siglech
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233274
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.