Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIGLEC12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110369
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110369 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110369 Supplier Invitrogen™ Supplier No. PA5110369
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EQKDYLLTMQ KSVTVQEGLC VSVLCSFSYP QNGWTASDPV HGYWFRAGDH VSRNIPVATN NPARAVQEET RDRFHLLGDP QNKDCTLSIR DTRESDAGTY VFCVERGNMK WNYKYDQLSV NVTASQDLLS RYRLEVPESV TVQEGLCVSV PCSVLYPHYN WTASSPVYGS WFKEGADIPW DIPVATNTPS G.

Sialic acid-binding immunoglobulin-like lectins are a family of cell surface proteins belonging to the immunoglobulin superfamily. They mediate protein-carbohydrate interactions by selectively binding to different sialic acid moieties present on glycolipids and glycoproteins. This gene encodes a member of the SIGLEC3-like subfamily of SIGLECs. Members of this subfamily are characterized by an extracellular V-set immunoglobulin-like domain followed by two C2-set immunoglobulin-like domains, and the cytoplasmic tyrosine-based motifs ITIM and SLAM-like. The encoded protein, upon tyrosine phosphorylation, has been shown to recruit the Src homology 2 domain-containing protein-tyrosine phosphatases SHP1 and SHP2. It has been suggested that the protein is involved in the negative regulation of macrophage signaling by functioning as an inhibitory receptor. This gene is located in a cluster with other SIGLEC3-like genes on 19q13.4. Alternatively spliced transcript variants encoding distinct isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIGLEC12
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Siglece
Gene Accession No. Q91Y57
Gene Alias Cd170; MIS; mSiglec-E; Myeloid inhibitory siglec; S2V; sialic acid binding Ig like lectin 12 (gene/pseudogene); sialic acid binding Ig-like lectin 12; sialic acid binding Ig-like lectin 5; sialic acid binding Ig-like lectin E; sialic acid-binding Ig-like lectin 12; Sialic acid-binding Ig-like lectin 5; sialic acid-binding Ig-like lectin E; Sialic acid-binding Ig-like lectin-like 1; sialic acid-binding immunoglobulin-like lectin E; Siglec; SIGLEC12; siglec-12; Siglec5; Siglec-5; Siglec9; Siglece; siglec-E; Siglecl1; Siglec-L1; SIGLEC-like 1; Siglec-XII; SLG; UNQ9215/PRO34042
Gene Symbols Siglece
Host Species Rabbit
Immunogen Recombinant protein (or fragment.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83382
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.