Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIGLEC15 Monoclonal Antibody (3C9C3)
GREENER_CHOICE

Catalog No. FERMA548818
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548818 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548818 Supplier Invitrogen™ Supplier No. MA548818
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SIGLEC15 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow, ELISA

Binds sialylated glycoproteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIGLEC15
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3C9C3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene SIGLEC15
Gene Accession No. Q6ZMC9
Gene Alias CD33 antigen-like 3; CD33 molecule-like 3; Cd33l3; EG620235; HsT1361; RGD1565948; sialic acid binding Ig like lectin 15; sialic acid binding Ig-like lectin 15; sialic acid-binding Ig-like lectin 15; Siglec; Siglec 15; SIGLEC15; Siglec-15; SIGLEC-I
Gene Symbols SIGLEC15
Host Species Mouse
Immunogen Purified recombinant fragment of human Siglec15 (AA: Extra(20-263)) expressed in Mammal.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284266
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.