Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIGLEC15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572765 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572765 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572765 Supplier Invitrogen™ Supplier No. PA572765
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

SIGLEC15 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Binds sialylated glycoproteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIGLEC15
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SIGLEC15
Gene Accession No. Q6ZMC9
Gene Alias CD33 antigen-like 3; CD33 molecule-like 3; Cd33l3; EG620235; HsT1361; RGD1565948; sialic acid binding Ig like lectin 15; sialic acid binding Ig-like lectin 15; sialic acid-binding Ig-like lectin 15; Siglec; Siglec 15; SIGLEC15; Siglec-15; SIGLEC-I
Gene Symbols SIGLEC15
Host Species Rabbit
Immunogen SIGLEC15 antibody for a 15 amino acid synthetic peptide near the amino terminus of human SIGLEC15. The immunogen is within amino acids 50 - 100 of SIGLEC15.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284266, 498888, 620235
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.