Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIGLEC5/SIGLEC14 Monoclonal Antibody (1A5), FITC
GREENER_CHOICE

Catalog No. PIMA528138
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528138 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528138 Supplier Invitrogen™ Supplier No. MA528138
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes human Siglec-5 (Sialic acid-binding Ig-like lectin 5), otherwise known as CD170, a novel sialic-acid-binding Ig-like lectin, and member of the Ig superfamily, expressed by dendritic cells (DCs), activated macrophages, neutrophils, and cells of the monocyte/myeloid lineage.

SIGLEC5 is a member of the sialic acid-binding immunoglobulin-like lectin (Siglec) family. These cell surface lectins are characterized by structural motifs in the immunoglobulin (Ig)-like domains and sialic acid recognition sites in the first Ig V set domain. The encoded protein is a member of the CD33-related subset of Siglecs and inhibits the activation of several cell types including monocytes, macrophages and neutrophils. Binding of group B Streptococcus (GBS) to the encoded protein plays a role in GBS immune evasion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIGLEC5/SIGLEC14
Applications Flow Cytometry
Classification Monoclonal
Clone 1A5
Concentration 100 μg/mL
Conjugate FITC
Formulation PBS with 1% BSA and 0.09% sodium azide
Gene SIGLEC5
Gene Accession No. O15389, Q08ET2
Gene Alias CD170; CD33 antigen-like 2; CD33L2; LLNLR-470E3.1; OB-binding protein 2; OBBP2; OB-BP2; obesity-binding protein 2; sialic acid binding Ig like lectin 14; sialic acid binding Ig like lectin 5; sialic acid binding Ig-like lectin 14; sialic acid binding Ig-like lectin 5; sialic acid-binding Ig-like lectin 14; Sialic acid-binding Ig-like lectin 5; sialic acid-binding immunoglobulin-like lectin 5; SIGLEC14; siglec-14; SIGLEC5; Siglec-5; UNQ294/PRO333
Gene Symbols SIGLEC14, SIGLEC5
Host Species Mouse
Immunogen Siglec-5-Fc protein, consisting of the full-length extracellular region of human Siglec-5, fused with the Fc region of human IgG1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100049587, 8778
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.