Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIGLEC9 Monoclonal Antibody (K8), PE

Catalog No. PIMA528202
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528202 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528202 Supplier Invitrogen™ Supplier No. MA528202

Mouse Monoclonal Antibody

Mouse anti Human Siglec-9, Clone K8, also known as CD329, recognizes Human sialic acid-binding Ig-like lectin 9, a single pass type I membrane protein belonging to the immunoglobulin superfamily. Reconstitute by adding 1 mL distilled water.

SIGLEC9 (Sialic acid-binding Ig-like lectin 9) is a putative adhesion molecule that mediates sialic-acid dependent binding to cells. It contains two Ig-like C2 type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. It preferentially binds to alpha2, 3-or alpha2, 6-linked sialic acid. The sialic acid recognition site may be mased by cis interactions with sialic acids on the same cell surface.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIGLEC9
Applications Flow Cytometry
Classification Monoclonal
Clone K8
Concentration 1 mg/mL
Conjugate PE
Formulation PBS with 1% BSA, 5% sucrose and 0.09% sodium azide
Gene SIGLEC9
Gene Accession No. Q9Y336
Gene Alias Cd170; CD329; CDw329; FOAP-9; LOC100062855; LOC719831; MIS; mSiglec-E; Myeloid inhibitory siglec; OBBP-LIKE; Protein FOAP-9; sialic acid binding Ig like lectin 9; sialic acid binding Ig-like lectin 5; sialic acid binding Ig-like lectin 9; sialic acid binding Ig-like lectin E; sialic acid-binding Ig-like lectin 12; Sialic acid-binding Ig-like lectin 5; sialic acid-binding Ig-like lectin 9; Sialic acid-binding Ig-like lectin E; sialic acid-binding Ig-like lectin-like 1; sialic acid-binding immunoglobulin-like lectin E; Siglec; Siglec12; siglec-12; Siglec5; Siglec-5; SIGLEC9; siglec-9; Siglece; Siglec-E; Siglecl1; siglec-L1; SIGLEC-like 1; UNQ668/PRO1302
Gene Symbols SIGLEC9
Host Species Mouse
Immunogen Recombinant Siglec-9 fused to Fc region of human IgG.
Purification Method Protein G
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27180
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.