Learn More
Invitrogen™ Sigma 54 Monoclonal Antibody (33.7)
Description
This antibody binds specifically in ELISA to monomeric tetrameric Sigma 54 but it does not bind to RAT protein or AgrA protein.
Specifications
Specifications
| Antigen | Sigma 54 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 33.7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Host Species | Mouse |
| Immunogen | Bacterially expressed Sigma 54 from Balb/c Sp2/0 cells, purified over a heparin column. |
| Purification Method | Protein G |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.