Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Diagnostics™ Silver Stain (Modified GMS) Kit

Catalog No. sdht100a
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SDHT100A 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SDHT100A Supplier Sigma Diagnostics™ Supplier No. HT100A1KT
Add to Cart
Add to Cart

Includes

  • Borax Solution (HT100-2); 100mL
  • Gold Chloride Solution (HT100-4); 100mL
  • Periodic Acid Solution (kit only); 100mL
  • Silver Methenamine Reagent (HT100-3); 12 vials
  • Sodium Thiosulfate Solution (HT100-5); 100mL

Intended for use in the histologic demonstration of fungi, basement membrane and some opportunistic organisms such as Pneumocystis carinii in tissue sections or smears

  • Incorporates a stable working silver methenamine salt along with buffer, toning reagent, and developer

For in vitro diagnostic use

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Distinguishing fungi, basement membrane and opportunistic organisms such as Pneumocystis carinii in tissue sections or smears.
Includes
  • Borax Solution (HT100-2); 100mL 
  • Gold Chloride Solution (HT100-4); 100mL
  • Periodic Acid Solution (kit only); 100mL
  • Silver Methenamine Reagent (HT100-3); 12 vials
  • Sodium Thiosulfate Solution (HT100-5); 100mL
Quantity 1 Kit
Storage Requirements 2° to 8°C
Synonym Modified Grocott Methanamine Silver (GMS) Fungus Stain, Periodic Acid Methanamine Silver Technique
Product Type Silver Stain (Modified GMS) Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.