Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Signature Series™ Bluing Reagent
GSA_VA

Catalog No. 22050115 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 gal.
1.32 gal.
4 x 1 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-050-115 4 x 1 gal.
22-050-114 1 gal.
73-01L 1.32 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-050-115 Supplier Epredia™ Supplier No. 7341
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Ensure crisp nuclear detail with the ready-to-use Epredia™ Signature Series Bluing Reagent.

  • Buffered to ensure proper alkalinity (pH = 8.0)
  • Specially formulated to prevent tissue separation from the slide
  • Requires only a one-minute water rinse versus conventional bluing reagents which require up-to five minutes

Available in 5 liter packaging

  • Reduces plastic by 90%
  • Increases stain stability
  • Reduces messy stain spills
  • Saves 35% storage space versus traditional packaging

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Bluing Reagent
Quantity 4 x 1 gal.
pH 8
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.