Learn More
Epredia™ Signature Series™ Clarifier™ 1, 2
GSA_VA
Catalog No. 22050118 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 gal.
1.32 gal.
4 x 1 gal.
Product Type:
Clarifier 1
Clarifier 2
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
22-050-118 4 x 1 gal. Clarifier 1
74-02L 1.32 gal. Clarifier 2
22-050-119 4 x 1 gal. Clarifier 2
22-050-116 1 gal. Clarifier 1
22-050-117 1 gal. Clarifier 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-050-118 Supplier Epredia™ Supplier No. 7441
Add to Cart
Add to Cart
This item is not returnable. View return policy
Enhance the nuclear definition and clarity of Signature Series hematoxylins with Epredia™ Signature Series Clarifier™ 1, 2.
  • Improves contrast between Nuclei and Cytoplasm
  • Increases cell transparency
  • Removes background staining caused by excessive tissue adhesive
  • Removes unwanted hematoxylin staining from adhesive without affecting nuclear staining
Note:
  • Clarifier 1 is recommended with Hematoxylin 7211 and Hematoxylin 1
  • Clarifier 2 is recommended with Hematoxylin 7212 and Hematoxylin 2
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 4 x 1 gal.
Product Type Clarifier 1
WARNING: Reproductive Harm - www.P65Warnings.ca.gov