Learn More
Epredia™ Signature Series™ Cyto-Stain™
Catalog No. p-4530166 Shop All Epredia Products
Change view
Click to view available options
Quantity:
4 pt.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-050-260 1 gal.
22-050-120 4 pt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-050-260 Supplier Epredia™ Supplier No. 7501R
Add to Cart
Add to Cart
This item is not returnable. View return policy
Achieve results similar to conventional Papanicolaou stains with Epredia™ Signature Series Cyto-Stain™, a uniquely formulated cytoplasmic stain
  • Use for all gynecological and nongynecological procedures
  • Reduces staining time
  • Cost-effective, as user performs only one counterstain
  • Cyto-Stain provides a full range of familiar cytoplasmic colors  and control over the staining intensities
  • Cyto-Stain G is a modification to produce greener cyanophilic hues in intermediate and basal cells
Specifications
Quantity 1 gal.
Product Type Cytoplasmic stain
WARNING: Reproductive Harm - www.P65Warnings.ca.gov