Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104295
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104295 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104295 Supplier Invitrogen™ Supplier No. PA5104295
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SIK2.

SNF1LK2 (QIK) is a serine/threonine kinase and is a salt-inducible kinase that belongs to the AMP-activated protein kinase family. SNF1LK2 negatively regulates CRE-binding protein (CREB) activity by phosphorylating the CREB-specific coactivator transducer of regulated CREB activity (TORC). SIK2 is thought to be part of a signaling cascade that regulates the expression and activity of the insulin-induced genes PGC-1'alpha and UCP-1 in brown adipocytes, impairment of which has been implicated in obesity and insulin resistance in human and animal models. SIK2 has also been reported as a key regulator for neuronal survival after ischemia, suppressing CREB-mediated gene expression after oxygen-glucose deprivation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIK2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SIK2
Gene Accession No. Q8CFH6, Q9H0K1
Gene Alias G630080D20Rik; KIAA0781; LOH11CR1I; QIK; Qin-induced kinase; RGD1559698; salt induceable kinase 2; salt inducible kinase 2; salt-inducible kinase 2; salt-inducible protein kinase 2; salt-inducible serine/threonine kinase 2; serine/threonine-protein kinase SIK2; Serine/threonine-protein kinase SNF1-like kinase 2; Sik2; SIK-2; SNF1-like kinase 2; SNF1LK2
Gene Symbols SIK2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SIK2(Accession Q9H0K1), corresponding to amino acid residues Y517-S567.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23235, 235344, 315649
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.