Learn More
Thermo Scientific™ SilTite™ Replacement Ferrules , Nuts and Baseplate Seals
Catalog No. 03050253 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Agilent Instruments
Agilent SSL Injectors
All SilTite Kits
MS Interface
Thermo Scientific DSQ GC-MS Interface Kits
Thermo Scientific TRACE 1300/1310 SSL and ISQ GC-MS Interface Kits
Includes:
Pk. of 10 Baseplate Seals
Pk. of 10 Ferrules
Pk. of 5 Nuts
Description:
290 MA201
290 MA202
290 MA203
290 MA205
290 MA207
290 MA228
290 MT221
290 MT222
290 MT223
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Includes Description
03-050-253 Agilent SSL Injectors Pk. of 5 Nuts 290 MA207
03-150-514 Thermo Scientific TRACE 1300/1310 SSL and ISQ GC-MS Interface Kits Pk. of 10 Ferrules 290 MA201
03-050-250 All SilTite Kits Pk. of 10 Ferrules 290 MA202
03-050-251 All SilTite Kits Pk. of 10 Ferrules 290 MA203
03-050-252 MS Interface Pk. of 5 Nuts 290 MA205
03-050-271 Thermo Scientific DSQ GC-MS Interface Kits Pk. of 10 Ferrules 290 MT221
03-050-272 Thermo Scientific DSQ GC-MS Interface Kits Pk. of 10 Ferrules 290 MT222
03-050-273 Thermo Scientific DSQ GC-MS Interface Kits Pk. of 10 Ferrules 290 MT223
03-050-259 Agilent Instruments Pk. of 10 Baseplate Seals 290 MA228
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 03-050-253 Supplier Thermo Scientific™ Supplier No. 290MA207
Add to Cart
Add to Cart
Eliminate leaks with the Thermo Scientific™ SilTite Ferrule.
Eliminate leaks with the Thermo Scientific™ SilTite Ferrule, which forms a strong, permanent, airtight seal around the capillary column.
  • SilTite metal ferrules and nuts are made from the same material and have the same thermal
  • expansion coefficient
  • Base of the ferrule is flat and forms a perfect seal with the MS interface
  • The ferrule's temperature tolerance is well above the limit of the injector, MS interface or GC oven
  • Unlike other ferrules, do not need re-tightening after installation
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Agilent SSL Injectors
Type SilTite Nuts
Description 290 MA207
Includes Pk. of 5 Nuts
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.