Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silica, fumed

Catalog No. 503126117 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-6117 100 g
50-312-6118 25 g
50-312-5936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-312-6117 Supplier Sigma Aldrich Supplier No. S5130100G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Fumed silica may be synthesized by high temperature hydrolysis of SiCl4 in O2(N2)/H2 flame. It is amorphous in nature and possesses very high specific area. The micro droplets of amorphous silica fuse into a branch and form a chain like agglomerate.Fumed silica is used to deplete lipids and hormones in Taqman assays of human hepatoma cells HuH-7 and human liver cell line HepG2. Preparation of epoxidized natural rubber/fumed silica composites have been reported. Interaction of Gemini surfactant C12-s-C122Br with aqueous suspension of fumed silica has been studied. Fumed silica based nanocomposites reinforced with organo clay may be prepared. TiO2/fumed silica porous ceramic material was used to prepare photocatalytic materials.Particle sizes are for primary particles, which form branched, chain-like aggregates a few tenths of a micron long.

Specifications

CAS 112945-52-5
Quantity 100 g
Molecular Formula O2Si
Linear Formula SiO2
MDL Number MFCD00011232
Synonym Silica; Silicic anhydride; Silicon dioxide; Silicon dioxide amorphous
Molecular Weight (g/mol) 60.08
Refractive Index n20/D 1.46 (literature)
Density 2.3 lb./cuft (at 25°C (bulk density)(literature))
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.