Learn More
MilliporeSigma™ Silica gel 60 GF254 for thin-layer chromatography, MilliporeSigma™
Catalog No. M1077300500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kg Plastic Bottle
25 kg Plastic Drum
5 kg Plastic Drum
500 g Plastic Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1077300500 500 g Plastic Bottle
M1077301000 1 kg Plastic Bottle
M1077305000 5 kg Plastic Drum
M1077309025 25 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M1077300500 Supplier MilliporeSigma™ Supplier No. 1.07730.0500
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Silica gel 60 GF254 for thin-layer chromatography. Silica gel 60 GF254 CAS No. 7631-86-9, EC Number 231-545-4.
Standardized sorbents ensure reliable results every time. Our broad spectrum meets all kinds of needs. In TLC, silica gel 60 sorbent is considered the most versatile. Our assortment of silica gel 60 features different grades with particle sizes ranging from 5-40μm: silica with a gypsum binder, silica with no foreign binder, and silica gel with a fluorescence indicator. We also offer high-quality aluminum oxide, cellulose microcrystalline, and kieselgur (also known as diatomaceous earth).
TRUSTED_SUSTAINABILITY
Specifications
Grade 60
Quantity 500 g Plastic Bottle
Type Packing Material
Material Silica Gel