Learn More
MilliporeSigma™ Silica gel 60 PF254 containing gypsum for preparative layer chromatography, MilliporeSigma™
Catalog No. M1077492500 Shop All MilliporeSigma Products
Change view
Click to view available options
Packaging:
Metal Can
Plastic Bottle
Plastic Drum
Quantity:
1 kg
2.5 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
M1077492500 Metal Can 2.5 kg
M1077499025 Plastic Drum 25 kg
M1077491000 Plastic Bottle 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1077492500 Supplier MilliporeSigma™ Supplier No. 1077492500
Add to Cart
Add to Cart
for preparative layer chromatography
Standardized sorbents ensure reliable results every time. Our broad spectrum meets all kinds of needs. In TLC, silica gel 60 sorbent is considered the most versatile. Our assortment of silica gel 60 features different grades with particle sizes ranging from 5-40μm: silica with a gypsum binder, silica with no foreign binder, and silica gel with a fluorescence indicator. We also offer high-quality aluminum oxide, cellulose microcrystalline, and kieselgur (also known as diatomaceous earth).
TRUSTED_SUSTAINABILITY
Specifications
Packaging Metal Can
Quantity 2.5 kg
Material Silica Gel 60 PF254 Containing Gypsum