Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica Gel, For Chromatography40-63 Micron (230-400 Mesh), 60A, Ultrapure, Spectrum™ Chemical
SDP

Catalog No. 18606202 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-202 2.5 kg Poly Pail
18-606-200 100 g Amber Glass Bottle
18-606-201 500 g Amber Glass Bottle
18-606-203 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-202 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SIL192.5KG
Add to Cart
Add to Cart

SIL19, 7631-86-9

Spectrum™ Chemical Silica Gel, For Chromatography40-63 Micron (230-400 Mesh), 60A, Ultrapure is a porous and granular form of silicon dioxide which is synthetically made from sodium silicate. In chemistry, the silica gel is used as a stationary phase in chromatography. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Specifications

Particle Size 40 to 63 μm
Grade Ultrapure
Packaging Poly Pail
pH 6.5 to 7.5
Quantity 2.5 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.