Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica Gel, Grade 633, 200-425 Mesh, 60A, Spectrum™ Chemical
SDP

Catalog No. 18606207 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
5 kg
25 kg
Packaging:
Steel Can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-207 25 kg Steel Can
18-606-204 100 g Steel Can
18-606-206 5 kg Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-606-207 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SIL6125KGBL
Add to Cart
Add to Cart

SIL61, 7631-86-9

Spectrum™ Chemical Silica Gel, Grade 633, 200-425 Mesh, 60A is a form of silicon dioxide made from sodium silicate. It is granular and porous solid and is a naturally occurring mineral. It is often used as a desiccant to control humidity and avoid spoilage. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

CAS Max % 1
Material Silica Gel
Quantity 25 kg
Packaging Steel Can
Pore Volume 0.70-0.85 CC/G
Grade Ungraded
Surface Area 480 m2/g
Chemical Name or Material Silica Gel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.