Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica gel, spherical, 100A, Thermo Scientific™

Catalog No. AA4409209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4409209 10 g
AA4409218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4409209 Supplier Thermo Scientific Chemicals Supplier No. 4409209

Useful product for column chromatography.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63231-67-4
Molecular Formula H2O3Si
Molecular Weight (g/mol) 78.098
MDL Number MFCD00011232
InChI Key IJKVHSBPTUYDLN-UHFFFAOYSA-N
Synonym metasilicic acid, silicic acid h2sio3, mikronisil, acidum silicicum, bio-sil, kieselsaure german, cubosic, dihydroxysilanone, silica acid, polysilicic acid
PubChem CID 14768
IUPAC Name dihydroxy(oxo)silane
SMILES O[Si](=O)O

Specifications

Quantity 10 g
Solubility Information Soluble in hydrofluoric acid. Insoluble in water.
Packaging Plastic bottle
Chemical Name or Material Silica gel
EINECSNumber 231-545-4
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.