Learn More
Sigma Organic Chemistry SILICA MESOPOROUS SBA-15 <150?M PARTICLE SIZE PORE SIZE 12 NM HEXAGONAL PORE MORPHOLOGY | 5G
Supplier:  Sigma Organic Chemistry 9138555G
Encompass_Preferred
SILICA MESOPOROUS SBA-15 <150?M PARTICLE SIZE PORE SIZE 12 NM HEXAGONAL PORE MORPHOLOGY, 5G
About This Item:
UNSPSC Code: 12352200
Storage: ROOM TEMPERATURE
Catalog No. 91-385-55G
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.