Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon diOxide, purum p.a., Acid purified, Solstice

Catalog No. 6002862 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
600-28-62 500 g
600-28-61 1 kg
600-28-63 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 600-28-62 Supplier Solstice Supplier No. 84880500G

Purum p.a., acid purified

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60676-86-0
Molecular Formula O2Si
Molecular Weight (g/mol) 60.083
MDL Number MFCD00011232
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Quantity 500 g
Linear Formula SiO2
UN Number NONH for all modes of transport
Formula Weight 60.08g/mol
Density 2.6g/mL at 25°C
Chemical Name or Material Silicon dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.