Learn More
Silicon(IV) nitride, Electronic Grade, 99.85% (metals basis), 90+% alpha-phase
Description
Silicon nitride bearings are both full ceramic bearings and ceramic hybrid bearings with balls in ceramics and races in steel. Silicon nitride ceramics have good shock resistance compared to other ceramics. Silicon nitride is used for rocker arm pads for lower wear, turbocharger for lower inertia and less engine lag, and in exhaust gas control valves for increased acceleration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1900°C (sublimation) |
| Color | Gray |
| Physical Form | Powder |
| Quantity | 10 g |
| Linear Formula | Si3N4 |
| Merck Index | 14,8497 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 140.28 |
| Percent Purity | 99.85% |
| Grade | Electronic |
| Show More |
Safety and Handling
| EINECSNumber | 234-796-8 |
| RTECSNumber | VW0650000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.