Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon(IV) oxide, 99.99% (metals basis)

Catalog No. AA4373614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4373614 25 g
AA4373622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4373614 Supplier Thermo Scientific Chemicals Supplier No. 04373614
Add to Cart

CAS: 7631-86-9 | O2Si | 60.08 g/mol

Silica is used primarily in the production of glass for windows, drinking glasses, beverage bottles. The majority of optical fibers for telecommunication are also made from silica. It is a primary raw material for many ceramics such as earthenware, stoneware, and porcelain. The main ingredient in sand casting for the manufacture of a large number of metallic components in engineering and other applications. Silica is a common additive in the production of foods. An estimated 95% of silicon dioxide produced is consumed in the construction industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Boiling Point 2230°C
Melting Point 1710°C
Color White
Physical Form Fused Lump
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 60.09
Odor Odorless
Density 2.2-2.6
Chemical Name or Material Silicon(IV) oxide
EINECSNumber 231-545-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.