Learn More
Silicon(IV) oxide, amorphous, nanopowder, 99.9% (metals basis)
Description
Silicon(IV) oxide is used as sodium aluminate stabilizer. It is also used as SDS mixture of sodium alkyl sulfates consisting chiefly of sodium lauryl sulfate. It is also acts as chain transfer agent in polymerization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 2230°C |
| Melting Point | 1710°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 100 g |
| Assay Percent Range | 99.9% (metals basis) |
| Solubility Information | Insoluble in water. |
| Formula Weight | 60.09 |
| Odor | Odorless |
| Density | 2.2 to 2.6 g/cm3 at 20°C |
| Show More |
Safety and Handling
| Hazard Category | H373 |
| Hazard Statement | Wear suitable protective clothing. |
| Precautionary Statement | P260-P314-P501c |
| EINECSNumber | 231-545-4 |
| RTECSNumber | VV7565000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.