Learn More
Silicon(IV) oxide, Amorphous fumed, S.A. 300 to 350m2/g
Description
Adhesives, coatings, cosmetics, defomers, inks, lubricants, sealants, thermal insulationSilicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of food and pharmaceutical products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 2230°C |
| Melting Point | 1710°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 50 g |
| Solubility Information | Insoluble in water. |
| IUPAC Name | dioxosilane |
| Formula Weight | 60.09 |
| Odor | Odorless |
| Density | 2.2 g/cm3 at 20°C |
| Show More |
Safety and Handling
| Hazard Category | H373: May cause damage to organs through prolonged or repeated exposure. |
| Hazard Statement | Wear suitable protective clothing. |
| EINECSNumber | 231-545-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.